4-Amino-1,5-dimethylimidazole hydrochloride 95%

Lieferant: Apollo Scientific

OR930171-100MG OR930171-1G OR930171-250MG
APOSOR930171-100MGEA 81.2 EUR
APOSOR930171-100MG APOSOR930171-1G APOSOR930171-250MG
4-Amino-1,5-dimethylimidazole hydrochloride 95%
4-Amino-1,5-dimethylimidazole hydrochloride
Formel: C₅H₉N₃·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD26383908
CAS-Nummer: 1456821-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR