6-tert-Butylnicotinic acid 98%

Lieferant: Apollo Scientific

Synonyme: 6-(tert-Butyl)nicotinic acid

OR930175-1G
APOSOR930175-1GEA 62.5 EUR
APOSOR930175-1G
6-tert-Butylnicotinic acid 98%
6-tert-Butylnicotinic acid
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08059362
CAS-Nummer: 832715-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR