2-Ethylindole 97%

Lieferant: Apollo Scientific

Synonyme: 2-Ethyl-1H-indole

OR930314-1G OR930314-5G
APOSOR930314-1GEA 106 EUR
APOSOR930314-1G APOSOR930314-5G
2-Ethylindole 97%
2-Ethylindole
Formel: C₁₀H₁₁N
Molecular Weight: 145,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00049343
CAS-Nummer: 3484-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR