tert-Butyl N-(1H-pyrazol-4-ylmethyl)carbamate 95%

Lieferant: Apollo Scientific

Synonyme: 4-(Boc-aminomethyl)pyrazole

OR930419-100MG OR930419-1G OR930419-250MG
APOSOR930419-100MGEA 130 EUR
APOSOR930419-100MG APOSOR930419-1G APOSOR930419-250MG
tert-Butyl N-(1H-pyrazol-4-ylmethyl)carbamate 95%
tert-Butyl N-(1H-pyrazol-4-ylmethyl)carbamate
Formel: C₉H₁₅N₃O₂
Molecular Weight: 197,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12820298
CAS-Nummer: 1107620-72-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR