5-Bromo-1H-indole-7-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR930421-1G
OR930421-250MG
APOSOR930421-1GEA
75.4
EUR
APOSOR930421-1G
APOSOR930421-250MG
5-Bromo-1H-indole-7-carboxylic acid 97%
5-Bromo-1H-indole-7-carboxylic acid
Formel:
C₉H₆BrNO₂ Molecular Weight: 240,053 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11845481 CAS-Nummer: 860624-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...