2-(Chloromethyl)pyridine 1-oxide 95%

Lieferant: Apollo Scientific

OR930466-1G OR930466-250MG
APOSOR930466-1GEA 688 EUR
APOSOR930466-1G APOSOR930466-250MG
2-(Chloromethyl)pyridine 1-oxide 95%
2-(Chloromethyl)pyridine 1-oxide
Formel: C₆H₆ClNO
Molecular Weight: 143,571 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10697575
CAS-Nummer: 31640-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR