2-Chloro-3-ethynylpyridine 95%

Lieferant: Apollo Scientific

OR930611-1G OR930611-250MG
APOSOR930611-1GEA 806 EUR
APOSOR930611-1G APOSOR930611-250MG
2-Chloro-3-ethynylpyridine 95%
2-Chloro-3-ethynylpyridine
Formel: C₇H₄ClN
Molecular Weight: 137.566 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189764
CAS-Nummer: 1196156-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR