2-(Fur-2-yl)benzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: [2-(Fur-2-yl)phenyl]methanol

OR9307-1G OR9307-250MG OR9307-5G
APOSOR9307-1GEA 149 EUR
APOSOR9307-1G APOSOR9307-250MG APOSOR9307-5G
2-(Fur-2-yl)benzyl alcohol 97%
2-(Fur-2-yl)benzyl alcohol
Formel: C₁₁H₁₀O₂
Molecular Weight: 174,196 g/mol
Schmelzpunkt: 85…86,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06203101
CAS-Nummer: 139697-88-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR