(4-Chloro-pyrimidin-2-yl)-cyclopropyl-amine 95%
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-N-cyclopropylpyrimidin-2-amine
OR930797-1G
APOSOR930797-1GEA
505
EUR
APOSOR930797-1G
(4-Chloro-pyrimidin-2-yl)-cyclopropyl-amine 95%
(4-Chloro-pyrimidin-2-yl)-cyclopropyl-amine
Formel:
C₇H₈ClN₃ Molecular Weight: 169,612 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11046853 CAS-Nummer: 1044771-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...