3-(Pyridin-3-yl)propan-1-amine 95%

Lieferant: Apollo Scientific

Synonyme: 3-(3-Pyridyl)propylamine

OR930832-1G OR930832-250MG
APOSOR930832-1GEA 229 EUR
APOSOR930832-1G APOSOR930832-250MG
3-(Pyridin-3-yl)propan-1-amine 95%
3-(Pyridin-3-yl)propan-1-amine
Formel: C₈H₁₂N₂
Molecular Weight: 136,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09886637
CAS-Nummer: 41038-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR