5-Amino-2-(pyrrolidin-1-yl)benzonitrile 95%
Lieferant: Apollo Scientific
OR930848-1G
OR930848-5G
APOSOR930848-1GEA
136
EUR
APOSOR930848-1G
APOSOR930848-5G
5-Amino-2-(pyrrolidin-1-yl)benzonitrile 95%
5-Amino-2-(pyrrolidin-1-yl)benzonitrile
Formel:
C₁₁H₁₃N₃ Molecular Weight: 187,241 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864592 CAS-Nummer: 219921-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...