1,2,3,4-Tetrahydro-4-methylquinoline 95%
Lieferant: Apollo Scientific
Synonyme:
4-Methyl-1,2,3,4-tetrahydroquinoline
OR930852-1G
OR930852-250MG
APOSOR930852-1GEA
140
EUR
APOSOR930852-1G
APOSOR930852-250MG
1,2,3,4-Tetrahydro-4-methylquinoline 95%
1,2,3,4-Tetrahydro-4-methylquinoline
Formel:
C₁₀H₁₃N Molecular Weight: 147,217 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047565 CAS-Nummer: 19343-78-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...