6-Methyl-1,2,3,4-tetrahydro-isoquinoline hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
6-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
OR931101-1G
OR931101-250MG
APOSOR931101-1GEA
335
EUR
APOSOR931101-1G
APOSOR931101-250MG
6-Methyl-1,2,3,4-tetrahydro-isoquinoline hydrochloride 95%
6-Methyl-1,2,3,4-tetrahydro-isoquinoline hydrochloride
Formel:
C₁₀H₁₃N·ClH Molecular Weight: 183,678 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832197 CAS-Nummer: 42923-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...