(2,3-Dihydro-1H-inden-2-yl)methanol 96%

Lieferant: Apollo Scientific

OR931116-1G OR931116-250MG
APOSOR931116-1GEA 531 EUR
APOSOR931116-1G APOSOR931116-250MG
(2,3-Dihydro-1H-inden-2-yl)methanol 96%
(2,3-Dihydro-1H-inden-2-yl)methanol
Formel: C₁₀H₁₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD08235017
CAS-Nummer: 5445-45-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR