4-Chloro-3-methyl-1H-pyrazole-5-carbaldehyde 95%
Lieferant: Apollo Scientific
OR931169-1G
OR931169-250MG
APOSOR931169-1GEA
712
EUR
APOSOR931169-1G
APOSOR931169-250MG
4-Chloro-3-methyl-1H-pyrazole-5-carbaldehyde 95%
4-Chloro-3-methyl-1H-pyrazole-5-carbaldehyde
Formel:
C₅H₅ClN₂O Molecular Weight: 144.559 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16556147 CAS-Nummer: 1050642-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...