(1R,2S,3R,4S)-1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-diol 95%
Lieferant: Apollo Scientific
OR931219-1G
OR931219-5G
APOSOR931219-1GEA
177
EUR
APOSOR931219-1G
APOSOR931219-5G
(1R,2S,3R,4S)-1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-diol 95%
(1R,2S,3R,4S)-1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-diol
Formel:
C₁₀H₁₈O₂ Molecular Weight: 170,249 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07784411 CAS-Nummer: 56614-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...