N-(1H-Indol-5-yl)acetamide 96%
Lieferant: Apollo Scientific
OR931441-1G
OR931441-250MG
APOSOR931441-1GEA
104
EUR
APOSOR931441-1G
APOSOR931441-250MG
N-(1H-Indol-5-yl)acetamide 96%
N-(1H-Indol-5-yl)acetamide
Formel:
C₁₀H₁₀N₂O Molecular Weight: 174,199 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01320495 CAS-Nummer: 7145-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...