2-Methyl-2H-indazole-4-carboxylic acid 95%
Lieferant: Apollo Scientific
OR931456-1G
OR931456-500MG
APOSOR931456-1GEA
864
EUR
APOSOR931456-1G
APOSOR931456-500MG
2-Methyl-2H-indazole-4-carboxylic acid 95%
2-Methyl-2H-indazole-4-carboxylic acid
Formel:
C₉H₈N₂O₂ Molecular Weight: 176,172 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15071443 CAS-Nummer: 1071433-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...