3-(2-Cyanopropan-2-yl)benzoic acid 95%

Lieferant: Apollo Scientific

OR931459-1G OR931459-250MG
APOSOR931459-1GEA 87 EUR
APOSOR931459-1G APOSOR931459-250MG
3-(2-Cyanopropan-2-yl)benzoic acid 95%
3-(2-Cyanopropan-2-yl)benzoic acid
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19694856
CAS-Nummer: 872091-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR