2-Chloro-5-cyanobenzene-1-sulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

OR931550-1G OR931550-250MG OR931550-5G
APOSOR931550-1GEA 108 EUR
APOSOR931550-1G APOSOR931550-250MG APOSOR931550-5G
2-Chloro-5-cyanobenzene-1-sulphonyl chloride 95%
2-Chloro-5-cyanobenzene-1-sulphonyl chloride
Formel: C₇H₃Cl₂NO₂S
Molecular Weight: 236.075 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18392427
CAS-Nummer: 942199-56-6
UN: 1759
ADR: 8 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR