8-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine 95%

Lieferant: Apollo Scientific

Synonyme: 8-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-amine

OR931617-1G OR931617-250MG
APOSOR931617-1GEA 142 EUR
APOSOR931617-1G APOSOR931617-250MG
8-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine 95%
8-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine
Formel: C₇H₈N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD11846609
CAS-Nummer: 1239648-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR