6-Isopropoxynicotinonitrile 95%
Lieferant: Apollo Scientific
OR931682-1G
OR931682-250MG
APOSOR931682-1GEA
59.6
EUR
APOSOR931682-1G
APOSOR931682-250MG
6-Isopropoxynicotinonitrile 95%
6-Isopropoxynicotinonitrile
Formel:
C₉H₁₀N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09930873 CAS-Nummer: 195140-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...