1-Methyl-1,2,3,4-tetrahydroquinoline-6-carbaldehyde 95%

Lieferant: Apollo Scientific

OR931708-1G OR931708-250MG
APOSOR931708-1GEA 159 EUR
APOSOR931708-1G APOSOR931708-250MG
1-Methyl-1,2,3,4-tetrahydroquinoline-6-carbaldehyde 95%
1-Methyl-1,2,3,4-tetrahydroquinoline-6-carbaldehyde
Formel: C₁₁H₁₃NO
Molecular Weight: 175,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03180300
CAS-Nummer: 493-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR