Methyl 4-amino-3-cyanobenzoate 95%

Lieferant: Apollo Scientific

OR931800-1G
APOSOR931800-1GEA 111 EUR
APOSOR931800-1G
Methyl 4-amino-3-cyanobenzoate 95%
Methyl 4-amino-3-cyanobenzoate
Formel: C₉H₈N₂O₂
Molecular Weight: 176.172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16038776
CAS-Nummer: 159847-80-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR