1-Allyl-1H-pyrrole-2-carbaldehyde 95%

Lieferant: Apollo Scientific

OR931806-1G OR931806-250MG
APOSOR931806-1GEA 400 EUR
APOSOR931806-1G APOSOR931806-250MG
1-Allyl-1H-pyrrole-2-carbaldehyde 95%
1-Allyl-1H-pyrrole-2-carbaldehyde
Formel: C₈H₉NO
Molecular Weight: 135.163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505317
CAS-Nummer: 101001-68-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR