2-(4-Aminobenzyl)aniline 98%
Lieferant: Apollo Scientific
Synonyme:
2-[(4-aminophenyl)methyl]aniline
OR931811-1G
OR931811-250MG
APOSOR931811-1GEA
287
EUR
APOSOR931811-1G
APOSOR931811-250MG
2-(4-Aminobenzyl)aniline 98%
2-(4-Aminobenzyl)aniline
Formel:
C₁₃H₁₄N₂ Molecular Weight: 198.264 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01670227 CAS-Nummer: 1208-52-2 UN: 2811 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...