tert-Butyl cyanomethylpropylcarbamate 95%

Lieferant: Apollo Scientific

OR931864-1G OR931864-250MG
APOSOR931864-1GEA 806 EUR
APOSOR931864-1G APOSOR931864-250MG
tert-Butyl cyanomethylpropylcarbamate 95%
tert-Butyl cyanomethylpropylcarbamate
Formel: C₁₀H₁₈N₂O₂
Molecular Weight: 198.262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09953056
CAS-Nummer: 1247796-07-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR