tert-Butylboronic acid, pinacol ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(tert-Butyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR9319-1G OR9319-5G
APOSOR9319-1GEA 21.9 EUR
APOSOR9319-1G APOSOR9319-5G
tert-Butylboronic acid, pinacol ester 97%
tert-Butylboronic acid, pinacol ester
Formel: C₁₀H₂₁BO₂
Molecular Weight: 184,084 g/mol
Dichte: 0,87 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD06657801
CAS-Nummer: 99810-76-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR