(2-Chloro-6-methyl-pyrimidin-4-yl)-cyclopropyl-amine 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-N-cyclopropyl-6-methylpyrimidin-4-amine

OR931917-1G
APOSOR931917-1GEA 517 EUR
APOSOR931917-1G
(2-Chloro-6-methyl-pyrimidin-4-yl)-cyclopropyl-amine 95%
(2-Chloro-6-methyl-pyrimidin-4-yl)-cyclopropyl-amine
Formel: C₈H₁₀ClN₃
Molecular Weight: 183,638 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16767268
CAS-Nummer: 1250038-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR