(6-Chloro-1-methyl-1H-benzo[d]imidazol-2-yl)methanol 95%

Lieferant: Apollo Scientific

OR932073-1G OR932073-250MG
APOSOR932073-1GEA 116 EUR
APOSOR932073-1G APOSOR932073-250MG
(6-Chloro-1-methyl-1H-benzo[d]imidazol-2-yl)methanol 95%
(6-Chloro-1-methyl-1H-benzo[d]imidazol-2-yl)methanol
Formel: C₉H₉ClN₂O
Molecular Weight: 196,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01073117
CAS-Nummer: 331949-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR