4-Chloro-7-hydroxyquinoline 95%

Lieferant: Apollo Scientific

OR932346-1G OR932346-250MG
APOSOR932346-1GEA 134 EUR
APOSOR932346-1G APOSOR932346-250MG
4-Chloro-7-hydroxyquinoline 95%
4-Chloro-7-hydroxyquinoline
Formel: C₉H₆ClNO
Molecular Weight: 179.603 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09261120
CAS-Nummer: 181950-57-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR