2-Biphenylacetic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-([1,1'-Biphenyl]-2-yl)acetic acid

OR932405-1G OR932405-250MG
APOSOR932405-1GEA 221 EUR
APOSOR932405-1G APOSOR932405-250MG
2-Biphenylacetic acid 97%
2-Biphenylacetic acid
Formel: C₁₄H₁₂O₂
Molecular Weight: 212,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06208460
CAS-Nummer: 14676-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR