Ethyl imidazo[1,2-b]pyridazine-2-carboxylate 95%

Lieferant: Apollo Scientific

OR932479-1G OR932479-250MG
APOSOR932479-1GEA 213 EUR
APOSOR932479-1G APOSOR932479-250MG
Ethyl imidazo[1,2-b]pyridazine-2-carboxylate 95%
Ethyl imidazo[1,2-b]pyridazine-2-carboxylate
Formel: C₉H₉N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD17016047
CAS-Nummer: 123531-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR