(4-Chloro-5-methyl-pyrimidin-2-yl)-cyclopropyl-amine 95%
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-N-cyclopropyl-5-methylpyrimidin-2-amine
OR932486-1G
APOSOR932486-1GEA
516
EUR
APOSOR932486-1G
(4-Chloro-5-methyl-pyrimidin-2-yl)-cyclopropyl-amine 95%
(4-Chloro-5-methyl-pyrimidin-2-yl)-cyclopropyl-amine
Formel:
C₈H₁₀ClN₃ Molecular Weight: 183,638 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18837194 CAS-Nummer: 1289385-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...