5-Isopropyl-4-methoxy-2-methylbenzaldehyde 95%
Lieferant: Apollo Scientific
Synonyme:
4-Methoxy-2-methyl-5-(1-methylethyl)benzaldehyde
OR932660-1G
OR932660-250MG
APOSOR932660-1GEA
134
EUR
APOSOR932660-1G
APOSOR932660-250MG
5-Isopropyl-4-methoxy-2-methylbenzaldehyde 95%
5-Isopropyl-4-methoxy-2-methylbenzaldehyde
Formel:
C₁₂H₁₆O₂ Molecular Weight: 192,254 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02629380 CAS-Nummer: 105337-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...