2-Methyl-2,3-dihydro-1-benzofuran-5-carbaldehyde 95%

Lieferant: Apollo Scientific

OR932719-1G OR932719-250MG
APOSOR932719-1GEA 712 EUR
APOSOR932719-1G APOSOR932719-250MG
2-Methyl-2,3-dihydro-1-benzofuran-5-carbaldehyde 95%
2-Methyl-2,3-dihydro-1-benzofuran-5-carbaldehyde
Formel: C₁₀H₁₀O₂
Molecular Weight: 162,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06255192
CAS-Nummer: 54365-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR