Methyl 2-(3-oxocyclobutyl)acetate

Lieferant: Apollo Scientific
Achtung

OR932724-1G OR932724-500MG
APOSOR932724-1GEA 1110 EUR
APOSOR932724-1G APOSOR932724-500MG
Methyl 2-(3-oxocyclobutyl)acetate
Methyl 2-(3-oxocyclobutyl)acetate
Formel: C₇H₁₀O₃
Molecular Weight: 142,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20134006
CAS-Nummer: 1148130-30-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR