4-Chloro-7-methoxyquinazoline 95%
Lieferant: Apollo Scientific
OR932740-1G
OR932740-250MG
APOSOR932740-1GEA
0
EUR
APOSOR932740-1G
APOSOR932740-250MG
4-Chloro-7-methoxyquinazoline 95%
4-Chloro-7-methoxyquinazoline
Formel:
C₉H₇ClN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD06738758 CAS-Nummer: 55496-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...