5-Bromo-1H-pyrazole-3-carboxylic acid 97%

Lieferant: Apollo Scientific

OR932764-1G OR932764-250MG
APOSOR932764-1GEA 114 EUR
APOSOR932764-1G APOSOR932764-250MG
5-Bromo-1H-pyrazole-3-carboxylic acid 97%
5-Bromo-1H-pyrazole-3-carboxylic acid
Formel: C₄H₃BrN₂O₂
Molecular Weight: 190.983 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21605341
CAS-Nummer: 1328893-16-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR