1H-[1,2,3]Triazolo[4,5-c]pyridine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3H-[1,2,3]Triazolo[4,5-c]pyridine

OR932797-1G OR932797-5G
APOSOR932797-1GEA 26.1 EUR
APOSOR932797-1G APOSOR932797-5G
1H-[1,2,3]Triazolo[4,5-c]pyridine 95%
1H-[1,2,3]Triazolo[4,5-c]pyridine
Formel: C₅H₄N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD02670924
CAS-Nummer: 273-05-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR