Methyl 2-(4-hydroxy-3-methylphenyl)acetate 95%
Lieferant: Apollo Scientific
OR932836-1G
OR932836-5G
APOSOR932836-1GEA
127
EUR
APOSOR932836-1G
APOSOR932836-5G
Methyl 2-(4-hydroxy-3-methylphenyl)acetate 95%
Methyl 2-(4-hydroxy-3-methylphenyl)acetate
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08234607 CAS-Nummer: 64360-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...