3-Chloro-5H-pyrrolo[2,3-b]pyrazine 98%

Lieferant: Apollo Scientific

OR932970-1G OR932970-250MG
APOSOR932970-1GEA 184 EUR
APOSOR932970-1G APOSOR932970-250MG
3-Chloro-5H-pyrrolo[2,3-b]pyrazine 98%
3-Chloro-5H-pyrrolo[2,3-b]pyrazine
Formel: C₆H₄ClN₃
Molecular Weight: 153,569 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12024539
CAS-Nummer: 1111638-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR