Methyl 2-amino-2-phenylpropanoate 98%

Lieferant: Apollo Scientific

OR933024-1G OR933024-250MG
APOSOR933024-1GEA 696 EUR
APOSOR933024-1G APOSOR933024-250MG
Methyl 2-amino-2-phenylpropanoate 98%
Methyl 2-amino-2-phenylpropanoate
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00268136
CAS-Nummer: 4507-41-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR