Ethyl 4-oxotetrahydrothiophene-3-carboxylate 95%
Lieferant: Apollo Scientific
OR933089-1G
OR933089-250MG
APOSOR933089-1GEA
88.6
EUR
APOSOR933089-1G
APOSOR933089-250MG
Ethyl 4-oxotetrahydrothiophene-3-carboxylate 95%
Ethyl 4-oxotetrahydrothiophene-3-carboxylate
Formel:
C₇H₁₀O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318533 CAS-Nummer: 78647-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...