2-(3,4-Dihydro-2H-quinolin-1-yl)-ethylamine 97%

Lieferant: Apollo Scientific

Synonyme: 2-(3,4-Dihydroquinolin-1(2H)-yl)ethanamine

OR933272-1G OR933272-5G
APOSOR933272-1GEA 348 EUR
APOSOR933272-1G APOSOR933272-5G
2-(3,4-Dihydro-2H-quinolin-1-yl)-ethylamine 97%
2-(3,4-Dihydro-2H-quinolin-1-yl)-ethylamine
Formel: C₁₁H₁₆N₂
Molecular Weight: 176,258 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738984
CAS-Nummer: 37481-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR