2-(1-Acetylpiperidin-2-yl)acetic acid 95%
Lieferant: Apollo Scientific
OR933389-1G
OR933389-250MG
APOSOR933389-1GEA
632
EUR
APOSOR933389-1G
APOSOR933389-250MG
2-(1-Acetylpiperidin-2-yl)acetic acid 95%
2-(1-Acetylpiperidin-2-yl)acetic acid
Formel:
C₉H₁₅NO₃ Molecular Weight: 185,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12784528 CAS-Nummer: 25393-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...