4,4-Dimethyl-2-(2-methylphenyl)-4,5-dihydro-1,3-oxazole 95%

Lieferant: Apollo Scientific

OR933391-1G OR933391-250MG
APOSOR933391-1GEA 790 EUR
APOSOR933391-1G APOSOR933391-250MG
4,4-Dimethyl-2-(2-methylphenyl)-4,5-dihydro-1,3-oxazole 95%
4,4-Dimethyl-2-(2-methylphenyl)-4,5-dihydro-1,3-oxazole
Formel: C₁₂H₁₅NO
Molecular Weight: 189,254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02234571
CAS-Nummer: 71885-44-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR