(3-Methylazetidin-3-yl)methanol hydrochloride
Lieferant: Apollo Scientific
OR934134-1G
OR934134-5G
APOSOR934134-1GEA
213
EUR
APOSOR934134-1G
APOSOR934134-5G
(3-Methylazetidin-3-yl)methanol hydrochloride
(3-Methylazetidin-3-yl)methanol hydrochloride
Formel:
C₅H₁₁NO·ClH Molecular Weight: 137.608 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28119098 CAS-Nummer: 1458653-12-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...