(R)-Octahydropyrazino[2,1-c][1,4]oxazine dihydrochloride 97%
Lieferant: Apollo Scientific
OR934263-1G OR934263-250MG
APOSOR934263-1GEA 702 EUR
APOSOR934263-1G APOSOR934263-250MG
(R)-Octahydropyrazino[2,1-c][1,4]oxazine dihydrochloride 97%
(R)-Octahydropyrazino[2,1-c][1,4]oxazine dihydrochloride
Formel: C₇H₁₄N₂O·2ClH
Molecular Weight: 215,121 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD27986913
CAS-Nummer: 1126432-04-9