2-(3-Oxocyclobutyl)acetic acid

Lieferant: Apollo Scientific

OR934535-1G OR934535-5G
APOSOR934535-1GEA 208 EUR
APOSOR934535-1G APOSOR934535-5G
2-(3-Oxocyclobutyl)acetic acid
2-(3-Oxocyclobutyl)acetic acid
Formel: C₆H₈O₃
Molecular Weight: 128,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28501618
CAS-Nummer: 1610028-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR